Please login to your online account to display your discounted pricing

Ethyl trifluoromethanesulfonate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC311850050

 View more versions of this product

Catalog No. AC311850050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 178.31
Quantity 5g
Density 1.3780g/mL
Chemical Name or Material Ethyl trifluoromethanesulfonate
Molecular Formula C3H5F3O3S
Linear Formula CF3SO3CH2CH3
CAS 425-75-2
Packaging Glass bottle
Assay Percent Range 98%
MDL Number MFCD00000410
Synonym Ethyl triflate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl trifluoromethanesulfonate
Assay Percent Range 98%
Molecular Formula C3H5F3O3S
Linear Formula CF3SO3CH2CH3
Formula Weight 178.31
MDL Number MFCD00000410
Synonym Ethyl triflate
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.

SDS & Safety Documents