missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl trifluoromethanesulfonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC311850010

 View more versions of this product

Catalog No. AC311850010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

178.31
1.3780g/mL
Ethyl trifluoromethanesulfonate
CF3SO3CH2CH3
MFCD00000410
Ethyl triflate
1g
98%
425-75-2
C3H5F3O3S
Glass bottle

Chemical Identifiers

98%
CF3SO3CH2CH3
MFCD00000410
Ethyl trifluoromethanesulfonate
C3H5F3O3S
178.31
Ethyl triflate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger