Learn More
Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1152018
Description
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.
Solubility
Slightly miscible with water. Miscible with chloroform and methanol.
Notes
Moisture Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Ethyl trifluoroacetate | |
<-78°C | |
60°C to 62°C | |
Sweet | |
1.307 | |
MFCD00000419 | |
UN2924 | |
Moisture sensitive | |
Slightly miscible with water. Miscible with chloroform and methanol. | |
CCOC(=O)C(F)(F)F | |
142.077 | |
142.08 |
383-63-1 | |
1.192 | |
−1°C (30°F) | |
C4H5F3O2 | |
CF3CO2CH2CH3 | |
50 g | |
1761411 | |
ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid | |
STSCVKRWJPWALQ-UHFFFAOYSA-N | |
ethyl 2,2,2-trifluoroacetate | |
9794 | |
99% |
Chemical Identifiers
383-63-1 | |
142.077 | |
STSCVKRWJPWALQ-UHFFFAOYSA-N | |
9794 | |
CCOC(=O)C(F)(F)F |
C4H5F3O2 | |
MFCD00000419 | |
ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid | |
ethyl 2,2,2-trifluoroacetate |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H225-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 206-851-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only