Please login to your online account to display your discounted pricing

Ethyl trans-3-ethoxyacrylate, 98%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224580025

 View more versions of this product

Catalog No. AC224580025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 144.17
Physical Form Liquid
Quantity 2.5g
Color Clear colorless to light yellow
Density 0.9900g/mL
Beilstein 03, 370
Linear Formula CH3CH2OCH=CHCOOCH2CH3
CAS 5941-55-9
Synonym Ethyl trans-3-ethoxy-2-propenoate
Packaging Glass bottle
Chemical Name or Material Ethyl trans-3-ethoxyacrylate
Assay Percent Range 98%
Molecular Formula C7H12O3
MDL Number MFCD00009863
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Ethyl trans-3-ethoxyacrylate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H12O3
Linear Formula CH3CH2OCH=CHCOOCH2CH3
Formula Weight 144.17
MDL Number MFCD00009863
Synonym Ethyl trans-3-ethoxy-2-propenoate
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents