missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trans-3-(1-pyrrolidinyl)acrylate, 98+%

$53.66 - $524.32

Chemical Identifiers

CAS 65651-80-1
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.224
MDL Number MFCD01075735
InChI Key VFFHLZJSZHWXAJ-VMPITWQZSA-N
Synonym ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
PubChem CID 639785
IUPAC Name ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate
SMILES CCOC(=O)C=CN1CCCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1636103 View Documents Alfa Aesar™
L1636103
1g Each for $53.66
 
AAL1636114 View Documents Alfa Aesar™
L1636114
25g Each for $524.32
 
AAL1636106 View Documents Alfa Aesar™
L1636106
5g Each for $155.76
 
Specifications

Chemical Identifiers

65651-80-1
169.224
VFFHLZJSZHWXAJ-VMPITWQZSA-N
639785
CCOC(=O)C=CN1CCCC1
C9H15NO2
MFCD01075735
ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate

Specifications

118°C (1mmHg)
1g
C9H15NO2
4399757
VFFHLZJSZHWXAJ-VMPITWQZSA-N
ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate
639785
≥98%
39°C to 43°C
65651-80-1
MFCD01075735
ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
CCOC(=O)C=CN1CCCC1
169.224
169.22
Ethyl trans-3-(1-pyrrolidinyl)acrylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C