Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trans-2-octenoate, 96%

$36.77 - $215.47

Chemical Identifiers

Chemical Name or Material Ethyl trans-2-octenoate
CAS 7367-82-0
Molecular Formula C10H18O2
Formula Weight 170.25
MDL Number MFCD00015287
Beilstein 1702980
Synonym trans-2-Octenoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1491906

  SDS
alfa aesar™
A1491906
5g Each for $36.77

AAA1491909

  SDS
alfa aesar™
A1491909
10g Each for $61.64

AAA1491918

  SDS
alfa aesar™
A1491918
50g Each for $215.47
Description & Specifications

Specifications

Formula Weight 170.25
Quantity 5g
Beilstein 1702980
Synonym trans-2-Octenoic acid ethyl ester
Boiling Point 110°C (10mmHg)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 1 Warning
Assay 96%
Molecular Formula C10H18O2
Chemical Name or Material Ethyl trans-2-octenoate
MDL Number MFCD00015287
Flash Point 91°C (195°F)
CAS 7367-82-0
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.