missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trans-2-octenoate, 96%

Manufacturer:  Alfa Aesar™ A1491906

 View more versions of this product

Catalog No. AAA1491906



Specifications

Specifications

Ethyl trans-2-octenoate
110°C (10mmHg)
7367-82-0
MFCD00015287
ethyl trans-2-octenoate, ethyl 2-octenoate, ethyl e-2-octenoate, ethyl e-oct-2-enoate, 2-octenoic acid, ethyl ester, ethyl 2-octenoate, trans, unii-44bpd919tx, 2-octenoic acid, ethyl ester, e, fema no. 3643, ethyl oct-2-enoate
CCCCCC=CC(=O)OCC
170.252
170.25
91°C (195°F)
5g
C10H18O2
1702980
AISZSTYLOVXFII-CMDGGOBGSA-N
ethyl (E)-oct-2-enoate
5364399
96%

Chemical Identifiers

7367-82-0
170.252
AISZSTYLOVXFII-CMDGGOBGSA-N
5364399
CCCCCC=CC(=O)OCC
C10H18O2
MFCD00015287
ethyl trans-2-octenoate, ethyl 2-octenoate, ethyl e-2-octenoate, ethyl e-oct-2-enoate, 2-octenoic acid, ethyl ester, ethyl 2-octenoate, trans, unii-44bpd919tx, 2-octenoic acid, ethyl ester, e, fema no. 3643, ethyl oct-2-enoate
ethyl (E)-oct-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents