missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trans-2-(hydroxymethyl)cyclopropanecarboxylate, 97%

Chemical Identifiers

CAS 1026787-30-3
Molecular Formula C7H12O3
MDL Number MFCD18434533
Synonym trans-2-(Hydroxymethyl)cyclopropanecarboxylic acid ethyl ester
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62920MD View Documents Alfa Aesar™
H62920MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1026787-30-3
C7H12O3
MFCD18434533
trans-2-(Hydroxymethyl)cyclopropanecarboxylic acid ethyl ester

Specifications

250mg
C7H12O3
trans-2-(Hydroxymethyl)cyclopropanecarboxylic acid ethyl ester
97%
1026787-30-3
MFCD18434533
144.16
Ethyl trans-2-(hydroxymethyl)cyclopropanecarboxylate