missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl trans-2-hexenoate, 98%

Manufacturer:  Alfa Aesar™ A1513618

 View more versions of this product

Catalog No. AAA1513618



Specifications

Specifications

Ethyl trans-2-hexenoate
62°C (25mmHg)
27829-72-7
MFCD00036541
1701323
SJRXWMQZUAOMRJ-VOTSOKGWSA-N
ethyl (E)-hex-2-enoate
5364778
98%
62°C (143°F)
50g
C8H14O2
UN3272
ethyl hex-2-enoate, ethyl 2-hexenoate, ethyl trans-2-hexenoate, 2-hexenoic acid, ethyl ester, ethyl e-hex-2-enoate, ethyl e-2-hexenoate, ethyl 2e-hexenoate, 2-hexenoic acid, ethyl ester, 2e, unii-ol4euz6c13, 2-hexenoic acid, ethyl ester, e
CCCC=CC(=O)OCC
142.198
142.2

Chemical Identifiers

27829-72-7
142.198
SJRXWMQZUAOMRJ-VOTSOKGWSA-N
5364778
CCCC=CC(=O)OCC
C8H14O2
MFCD00036541
ethyl hex-2-enoate, ethyl 2-hexenoate, ethyl trans-2-hexenoate, 2-hexenoic acid, ethyl ester, ethyl e-hex-2-enoate, ethyl e-2-hexenoate, ethyl 2e-hexenoate, 2-hexenoic acid, ethyl ester, 2e, unii-ol4euz6c13, 2-hexenoic acid, ethyl ester, e
ethyl (E)-hex-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Documents