Please login to your online account to display your discounted pricing

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  265652500

 View more versions of this product

Catalog No. AC265652500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (Argentometry)
Physical Form Granular Powder
Formula Weight 207.7
Quantity 250mg
Color White to almost white
Melting Point 152° to 156°C
Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Molecular Formula C9H17NO2·HCl
CAS 28250-14-8
Infrared Spectrum Authentic
Assay Percent Range 98%

Specifications

Chemical Name or Material Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C9H17NO2·HCl
Formula Weight 207.7