missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 28250-14-8 |
---|---|
Molecular Formula | C9H17NO2·HCl |
Molecular Weight (g/mol) | 207.7 |
InChI Key | XMQSOBPCWYVZSW-SCLLHFNJSA-N |
Synonym | ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, |
PubChem CID | 2724642 |
IUPAC Name | ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride |
SMILES | CCOC(=O)C1CCCCC1N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC265650010
|
Thermo Scientific Chemicals
265650010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
28250-14-8 | |
207.7 | |
ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, | |
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride |
C9H17NO2·HCl | |
XMQSOBPCWYVZSW-SCLLHFNJSA-N | |
2724642 | |
CCOC(=O)C1CCCCC1N.Cl |
Specifications
28250-14-8 | |
97.5% min. (Argentometry) | |
C9H17NO2·HCl | |
ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, 1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, | |
XMQSOBPCWYVZSW-SCLLHFNJSA-N | |
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride | |
2724642 | |
98% |
Authentic | |
Glass bottle | |
1g | |
Solubility in water: soluble in water | |
CCOC(=O)C1CCCCC1N.Cl | |
207.7 | |
207.7 | |
Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride |
RUO â Research Use Only