missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC265650050

 View more versions of this product

Catalog No. AC265650050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride
Glass Bottle
152.0°C to 156.0°C
28250-14-8
C9H17NO2·HCl
Solubility in water: soluble in water
CCOC(=O)C1CCCCC1N.Cl
207.7
207.7
98%
Authentic
White
5g
97.5% min. (Argentometry)
1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride,, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride
XMQSOBPCWYVZSW-SCLLHFNJSA-N
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride
2724642
Granular Powder

Chemical Identifiers

28250-14-8
207.7
1r,2r-ethyl 2-aminocyclohexanecarboxylate hydrochloride, ethyl 1r,2r-2-aminocyclohexane-1-carboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride,, trans-2-amino-cyclohexanecarboxylic acid ethyl ester hydrochloride, trans-ethyl 2-aminocyclohexanecarboxylate hydrochloride
ethyl (1R,2R)-2-aminocyclohexane-1-carboxylate;hydrochloride
C9H17NO2·HCl
XMQSOBPCWYVZSW-SCLLHFNJSA-N
2724642
CCOC(=O)C1CCCCC1N.Cl
SDS Documents