Learn More
Ethyl tiglate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1202914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl tiglate | |
0.93 | |
46°C (114°F) | |
1.434 | |
MFCD00015183 | |
UN3272 | |
14,9433 | |
OAPHLAAOJMTMLY-GQCTYLIASA-N | |
ethyl (E)-2-methylbut-2-enoate | |
5281163 | |
128.17 |
5837-78-5 | |
156°C | |
C7H12O2 | |
CH3CH=C(CH3)CO2CH2CH3 | |
25 g | |
1720895 | |
ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate | |
CCOC(=O)C(=CC)C | |
128.171 | |
CHEBI:4892 | |
98% |
Chemical Identifiers
5837-78-5 | |
128.171 | |
OAPHLAAOJMTMLY-GQCTYLIASA-N | |
5281163 | |
ethyl (E)-2-methylbut-2-enoate |
C7H12O2 | |
MFCD00015183 | |
ethyl tiglate, tiglic acid, ethyl ester, ethyl 2-methylcrotonate, ethyl alpha-methylcrotonate, ethyl trans-2-methyl-2-butenoate, e-2-methyl-2-butenoic acid ethyl ester, ethyl e-2-methylcrotonate, ethyl trans-2-methylcrotonate, tiglic acid ethyl ester, ethyl 2-methylbut-2-enoate | |
CHEBI:4892 | |
CCOC(=O)C(=CC)C |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 227-425-6
RTECSNumber : EM9252700
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only