missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2246814
Description
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl thiophene-3-carboxylate is used as an intermediate in organic syntheses
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Ethyl thiophene-3-carboxylate | |
207°C to 208°C | |
1.5225 | |
25 g | |
Soluble in water. | |
CCOC(=O)C1=CSC=C1 | |
156.199 | |
156.2 |
5751-80-4 | |
C7H8O2S | |
MFCD02179226 | |
ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n | |
OYSLMAQEMAJMCL-UHFFFAOYSA-N | |
ethyl thiophene-3-carboxylate | |
79824 | |
98% |
Chemical Identifiers
5751-80-4 | |
156.199 | |
OYSLMAQEMAJMCL-UHFFFAOYSA-N | |
79824 | |
CCOC(=O)C1=CSC=C1 |
C7H8O2S | |
MFCD02179226 | |
ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n | |
ethyl thiophene-3-carboxylate |
Safety and Handling
EINECSNumber : 227-275-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only