Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl thiophene-2-carboxylate, 98+%

$126.53 - $126.53

Chemical Identifiers

Chemical Name or Material Ethyl thiophene-2-carboxylate
CAS 2810-04-0
Molecular Formula C7H8O2S
Formula Weight 156.2
MDL Number MFCD00005436
Beilstein 112678
Synonym Thiophene-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1230218

 
alfa aesar™
A1230218
50g Each for $126.53
Description & Specifications

Specifications

Formula Weight 156.2
Chemical Name or Material Ethyl thiophene-2-carboxylate
Assay ≥98%
Molecular Formula C7H8O2S
MDL Number MFCD00005436
CAS 2810-04-0
Beilstein 112678
Synonym Thiophene-2-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 217° to 218°C
Quantity 50g
Flash Point 88°C (190°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.