Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl thiooxamate, 95%

$52.03 - $566.46

Chemical Identifiers

Chemical Name or Material Ethyl thiooxamate
CAS 16982-21-1
Molecular Formula C4H7NO2S
Formula Weight 133.17
MDL Number MFCD00074903
Synonym Ethyl aminothioxoacetate; Thiooxamic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3144703

 
alfa aesar™
H3144703
1g Each for $52.03

AAH3144706

 
alfa aesar™
H3144706
5g Each for $172.41

AAH3144714

 
alfa aesar™
H3144714
25g Each for $566.46
Description & Specifications

Specifications

Formula Weight 133.17
Chemical Name or Material Ethyl thiooxamate
Assay 95%
Molecular Formula C4H7NO2S
MDL Number MFCD00074903
CAS 16982-21-1
Synonym Ethyl aminothioxoacetate; Thiooxamic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 61° to 65°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.