missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl thiooxamate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3144706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl thiooxamate | |
61°C to 65°C | |
MFCD00074903 | |
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester | |
CCOC(=O)C(=S)N | |
133.165 | |
133.17 |
16982-21-1 | |
C4H7NO2S | |
5 g | |
YMBMCMOZIGSBOA-UHFFFAOYSA-N | |
ethyl 2-amino-2-sulfanylideneacetate | |
2733398 | |
95% |
Chemical Identifiers
16982-21-1 | |
133.165 | |
YMBMCMOZIGSBOA-UHFFFAOYSA-N | |
2733398 | |
CCOC(=O)C(=S)N |
C4H7NO2S | |
MFCD00074903 | |
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester | |
ethyl 2-amino-2-sulfanylideneacetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 605-564-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only