Please login to your online account to display your discounted pricing

Ethyl tetrahydropyran-4-ylacetate, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC67423DA

 View more versions of this product

Catalog No. CC67423DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 172.22
Quantity 1g
Assay 97%
CAS 103260-44-2
Packaging Amber Glass Bottle
Chemical Name or Material Ethyl tetrahydropyran-4-ylacetate
Assay Percent Range 97%
Molecular Formula C9H16O3

Specifications

Chemical Name or Material Ethyl tetrahydropyran-4-ylacetate
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16O3
Formula Weight 172.22