missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Ethyl tetrahydropyran-4-ylacetate, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
103260-44-2 |
Molecular Formula |
C9H16O3 |
Molecular Weight (g/mol) |
172.224 |
InChI Key |
JLMMMEDWRUVCEW-UHFFFAOYSA-N |
Synonym |
ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate |
PubChem CID |
2773412 |
IUPAC Name |
ethyl 2-(oxan-4-yl)acetate |
SMILES |
CCOC(=O)CC1CCOCC1 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
103260-44-2 |
|
172.224 |
|
ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate |
|
ethyl 2-(oxan-4-yl)acetate |
|
C9H16O3 |
|
JLMMMEDWRUVCEW-UHFFFAOYSA-N |
|
2773412 |
|
CCOC(=O)CC1CCOCC1 |
Specifications
|
103260-44-2 |
|
Amber Glass Bottle |
|
1g |
|
JLMMMEDWRUVCEW-UHFFFAOYSA-N |
|
ethyl 2-(oxan-4-yl)acetate |
|
2773412 |
|
97% |
|
97% |
|
C9H16O3 |
|
ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate |
|
CCOC(=O)CC1CCOCC1 |
|
172.224 |
|
172.22 |
|
Ethyl tetrahydropyran-4-ylacetate |
SDS
missing translation for 'documents'