missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl tetradecanoate, 98%

Manufacturer:  Alfa Aesar™ A11646AE

 View more versions of this product

Catalog No. AAA11646AE



Description

Description

Applications
A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications

Specifications

Ethyl tetradecanoate
Weak
12°C
124-06-1
CH3(CH2)12CO2CH2CH3
1776382
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
CCCCCCCCCCCCCC(=O)OCC
256.43
CHEBI:84849
98%
135°C (275°F)
295°C
100mL
C16H32O2
MFCD00008984
14,6333
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl tetradecanoate
31283
256.43

Chemical Identifiers

124-06-1
256.43
MMKRHZKQPFCLLS-UHFFFAOYSA-N
31283
ethyl tetradecanoate
C16H32O2
MFCD00008984
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
CHEBI:84849
CCCCCCCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents