missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl tert-Butylacetate, TCI America™

$129.86 - $385.02

Chemical Identifiers

CAS 5340-78-3
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00026911
InChI Key JWMNHAMYTBAUPI-UHFFFAOYSA-N
Synonym ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
PubChem CID 79284
IUPAC Name ethyl 3,3-dimethylbutanoate
SMILES CCOC(=O)CC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B151025ML View Documents TCI America
B151025ML
25mL Each for $129.86
 
B1510100ML View Documents TCI America
B1510100ML
100mL Each for $385.02
 
Specifications

Chemical Identifiers

5340-78-3
144.214
JWMNHAMYTBAUPI-UHFFFAOYSA-N
79284
CCOC(=O)CC(C)(C)C
C8H16O2
MFCD00026911
ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
ethyl 3,3-dimethylbutanoate

Specifications

145°C
25mL
C8H16O2
3272
JWMNHAMYTBAUPI-UHFFFAOYSA-N
ethyl 3,3-dimethylbutanoate
79284
Clear Liquid at 20°C
Ethyl tert-Butylacetate
Colorless
5340-78-3
MFCD00026911
ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
CCOC(=O)CC(C)(C)C
144.214
144.21
≥99.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0782

TSCA : No