missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl tert-Butylacetate 99.0+%, TCI America™
Supplier: TCI America B1510100ML
Specifications
Ethyl tert-Butylacetate | |
Colorless | |
C8H16O2 | |
100 mL | |
ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester | |
CCOC(=O)CC(C)(C)C | |
144.214 | |
144.21 | |
Liquid |
5340-78-3 | |
145°C | |
MFCD00026911 | |
3272 | |
JWMNHAMYTBAUPI-UHFFFAOYSA-N | |
ethyl 3,3-dimethylbutanoate | |
79284 | |
≥99.0% (GC) |
Chemical Identifiers
5340-78-3 | |
144.214 | |
JWMNHAMYTBAUPI-UHFFFAOYSA-N | |
79284 | |
CCOC(=O)CC(C)(C)C |
C8H16O2 | |
MFCD00026911 | |
ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester | |
ethyl 3,3-dimethylbutanoate |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : No