missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Sulfide 98.0+%, TCI America™

Supplier:  TCI America E0176100ML

 View more versions of this product

Catalog No. E0176100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Sulfide
Colorless
C4H10S
100 mL
diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
CCSCC
90.18
CHEBI:27710
≥98.0% (GC)
352-93-2
92°C
MFCD00009270
2375
LJSQFQKUNVCTIA-UHFFFAOYSA-N
(ethylsulfanyl)ethane
9609
90.18
Liquid

Chemical Identifiers

352-93-2
90.18
LJSQFQKUNVCTIA-UHFFFAOYSA-N
9609
(ethylsulfanyl)ethane
C4H10S
MFCD00009270
diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
CHEBI:27710
CCSCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LC7200000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions