Please login to your online account to display your discounted pricing

Ethyl succinyl chloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  184090250

 View more versions of this product

Catalog No. AC184090250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay 97.5 to 102.5% (RCOCl), ≥97.5% (GC)
Formula Weight 164.59
Quantity 25g
Color Clear colorless to yellow
Beilstein 02, 613
EINECS Number 238-855-9
TSCA TSCA
Specific Gravity 1.144 to 1.17 (20°C)
Density 1.1575g/mL
Refractive Index 1.4350 to 1.4390 (20°C, 589nm)
Synonym Ethyl 3-(chloroformyl)propionate
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Packaging Glass bottle
Chemical Name or Material Ethyl succinyl chloride
Linear Formula ClCOCH2CH2CO2C2H5
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H9ClO3
MDL Number MFCD00000751
CAS 14794-31-1

Specifications

Chemical Name or Material Ethyl succinyl chloride
Name Note 98%
CAS 14794-31-1
Assay 97.5 to 102.5% (RCOCl), ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H9ClO3
Linear Formula ClCOCH2CH2CO2C2H5
Formula Weight 164.59
MDL Number MFCD00000751
Synonym Ethyl 3-(chloroformyl)propionate
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement Causes severe skin burns and eye damage.

SDS & Safety Documents