Learn More
Ethyl succinyl chloride, 95%, Thermo Scientific Chemicals
$117.47 - $1324.40
Chemical Identifiers
CAS | 14794-31-1 |
---|---|
Molecular Formula | C6H9ClO3 |
Molecular Weight (g/mol) | 164.585 |
MDL Number | MFCD00000751 |
InChI Key | IXZFDJXHLQQSGQ-UHFFFAOYSA-N |
Synonym | ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride |
PubChem CID | 84648 |
IUPAC Name | ethyl 4-chloro-4-oxobutanoate |
SMILES | CCOC(=O)CCC(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1273414
|
Thermo Scientific Chemicals
A1273414 |
25 g |
Each for $117.47
|
|
|||||
AAA1273422
|
Thermo Scientific Chemicals
A1273422 |
100 g |
Each for $342.49
|
|
|||||
AAA1273436
|
Thermo Scientific Chemicals
A1273436 |
500 g |
Each for $1,324.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14794-31-1 | |
164.585 | |
IXZFDJXHLQQSGQ-UHFFFAOYSA-N | |
84648 | |
CCOC(=O)CCC(=O)Cl |
C6H9ClO3 | |
MFCD00000751 | |
ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride | |
ethyl 4-chloro-4-oxobutanoate |
Specifications
14794-31-1 | |
88°C to 90°C (11 mmHg) | |
Acrid | |
1.437 | |
25 g | |
1766126 | |
ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride | |
CCOC(=O)CCC(=O)Cl | |
164.585 | |
164.59 | |
Ethyl succinyl chloride |
1.163 | |
84°C (183°F) | |
C6H9ClO3 | |
MFCD00000751 | |
UN3265 | |
Moisture sensitive | |
IXZFDJXHLQQSGQ-UHFFFAOYSA-N | |
ethyl 4-chloro-4-oxobutanoate | |
84648 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 238-855-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only