missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Stilbene-4-carboxylate 98.0+%, TCI America™
Supplier: TCI America E06785G
Specifications
Ethyl Stilbene-4-carboxylate | |
110°C | |
MFCD06204340 | |
Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene | |
CCOC(=O)C1=CC=C(C=C1)C=CC2=CC=CC=C2 | |
252.313 | |
252.31 | |
Crystalline Powder |
109463-48-1 | |
C17H16O2 | |
5 g | |
RBQGAPDZGPQIDL-CMDGGOBGSA-N | |
ethyl 4-[(E)-2-phenylethenyl]benzoate | |
5907907 | |
≥98.0% (GC) |
Chemical Identifiers
109463-48-1 | |
252.313 | |
RBQGAPDZGPQIDL-CMDGGOBGSA-N | |
5907907 | |
CCOC(=O)C1=CC=C(C=C1)C=CC2=CC=CC=C2 |
C17H16O2 | |
MFCD06204340 | |
Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene | |
ethyl 4-[(E)-2-phenylethenyl]benzoate |
Safety and Handling
TSCA : Yes