missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Sorbate 98.0+%, TCI America™
Supplier: TCI America S0055100ML
Specifications
| Ethyl Sorbate | |
| Yellow | |
| C8H12O2 | |
| 100 mL | |
| OZZYKXXGCOLLLO-TWTPFVCWSA-N | |
| ethyl (2E,4E)-hexa-2,4-dienoate | |
| 1550470 | |
| ≥98.0% (GC) |
| 2396-84-1 | |
| 76°C | |
| MFCD00009296 | |
| ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459 | |
| CCOC(=O)\C=C\C=C\C | |
| 140.18 | |
| 140.18 | |
| Liquid |
Chemical Identifiers
| 2396-84-1 | |
| 140.18 | |
| OZZYKXXGCOLLLO-TWTPFVCWSA-N | |
| 1550470 | |
| CCOC(=O)\C=C\C=C\C |
| C8H12O2 | |
| MFCD00009296 | |
| ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459 | |
| ethyl (2E,4E)-hexa-2,4-dienoate |
Safety and Handling
EINECSNumber : (2)-1081
RTECSNumber : WG2143000
TSCA : Yes