Please login to your online account to display your discounted pricing

Ethyl sorbate, 97%, stabilized, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC365420250

 View more versions of this product

Catalog No. AC365420250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 140.18
Quantity 25g
Color Clear colorless to yellow
Beilstein 02, 484
Density 0.933g/mL
Packaging Glass bottle
Chemical Name or Material Ethyl sorbate
Refractive Index 1.4930 to 1.4960 (20°C, 589nm)
Stabilizer 500ppm DL-alpha-Tocopherole
Acid Value 1mg KOH/g max.
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H12O2
Linear Formula CH3CH=CHCH=CHCOCH5
MDL Number MFCD00009296
CAS 2396-84-1
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Ethyl sorbate
Name Note 97%
CAS 2396-84-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H12O2
Linear Formula CH3CH=CHCH=CHCOCH5
Formula Weight 140.18
MDL Number MFCD00009296