Please login to your online account to display your discounted pricing

Ethyl sorbate, 97%, stabilized, ACROS Organics™

$27.81 - $27.81

Chemical Identifiers

Chemical Name or Material Ethyl sorbate
Name Note 97%
CAS 2396-84-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H12O2
Linear Formula CH3CH=CHCH=CHCOCH5
Formula Weight 140.18
MDL Number MFCD00009296
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC365420250

  SDS
fisher scientific acros
AC365420250
Glass bottle 25g Each for $27.81
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 140.18
Physical Form Liquid
Quantity 25g
Color Clear colorless to yellow
Beilstein 02, 484
Density 0.933g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 2396-84-1
Linear Formula CH3CH=CHCH=CHCOCH5
Refractive Index 1.4930 to 1.4960 (20°C, 589nm)
Acid Value 1mg KOH/g max.
Stabilizer 500ppm DL-alpha-Tocopherole
Chemical Name or Material Ethyl sorbate
Molecular Formula C8H12O2
Name Note 97%
MDL Number MFCD00009296
Assay Percent Range 97%