missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl salicylate, 99%

Manufacturer:  Alfa Aesar™ A101010E

 View more versions of this product

Catalog No. AAA101010E



Specifications

Specifications

Ethyl salicylate
232°C
2500g
C9H10O3
907659
ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
CCOC(=O)C1=CC=CC=C1O
166.176
166.18
107°C (224°F)
1°C
118-61-6
MFCD00002215
14,3850
GYCKQBWUSACYIF-UHFFFAOYSA-N
ethyl 2-hydroxybenzoate
8365
99%

Chemical Identifiers

118-61-6
166.176
GYCKQBWUSACYIF-UHFFFAOYSA-N
8365
CCOC(=O)C1=CC=CC=C1O
C9H10O3
MFCD00002215
ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
ethyl 2-hydroxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VO3000000

TSCA : TSCA

SDS Documents