Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (S)-indoline-2-carboxylate hydrochloride, 98%

$56.96 - $135.15

Chemical Identifiers

Chemical Name or Material Ethyl (S)-indoline-2-carboxylate hydrochloride
CAS 79854-42-5
Molecular Formula C11H13NO2·HCl
Formula Weight 227.69
MDL Number MFCD09033207
Synonym (S)-Indoline-2-carboxylic acid ethyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2805903

  SDS
alfa aesar™
H2805903
1g Each for $135.15

AAH28059MD

  SDS
alfa aesar™
H28059MD
250mg Each for $56.96
Description & Specifications

Specifications

Formula Weight 227.69
Chemical Name or Material Ethyl (S)-indoline-2-carboxylate hydrochloride
Assay 98%
Molecular Formula C11H13NO2·HCl
MDL Number MFCD09033207
CAS 79854-42-5
Synonym (S)-Indoline-2-carboxylic acid ethyl ester hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 152° to 158°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.