Please login to your online account to display your discounted pricing

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  361290050

 View more versions of this product

Catalog No. AC361290050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 166.61
Quantity 5g
Color Clear colorless to yellow
Density 1.1900g/mL
Specific Rotation Condition − 14.50 (20.00°C c=neat)
Refractive Index 1.4515 to 1.4535 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Specific Rotation -13° to -16° (20°C, 589nm) (neat)
Packaging Glass bottle
Molecular Formula C6H11ClO3
Chemical Name or Material Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
MDL Number MFCD00211241
Assay Percent Range 97%
CAS 86728-85-0

Specifications

Chemical Name or Material Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
CAS 86728-85-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H11ClO3
Formula Weight 166.61
MDL Number MFCD00211241
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.

SDS & Safety Documents