missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 97%, ACROS Organics™

Chemical Identifiers

CAS 86728-85-0
Assay Percent Range 97%
Molecular Formula C6H11ClO3
Formula Weight 166.61
MDL Number MFCD00211241
Chemical Name or Material Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC361290010 SDS Thermo Fisher Scientific
361290010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC361290050 SDS Thermo Fisher Scientific
361290050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

86728-85-0
C6H11ClO3
MFCD00211241
≥96.0%
97%
166.61
Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate

Specifications

1g
1.1900g/mL
Glass bottle
-13° to -16° (20°C, 589nm) (neat)
Clear colorless to yellow
97%
166.61
Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
86728-85-0
Authentic
1.4515 to 1.4535 (20°C, 589nm)
− 14.50 (20.00°C c=neat)
Liquid
C6H11ClO3
MFCD00211241
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.