missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (S)-3-Piperidinecarboxylate D-Tartrate, TCI America™

$157.36 - $537.78

Chemical Identifiers

CAS 83602-38-4
Molecular Formula C12H21NO8
Molecular Weight (g/mol) 307.299
MDL Number MFCD03093639
InChI Key HHPGQKZOPPDLNH-FWQMQHHGSA-N
Synonym (S)-Nipecotic Acid Ethyl Ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, (S)-3-Piperidinecarboxylic Acid Ethyl Ester D-Tartrate
PubChem CID 118855422
IUPAC Name (3S)-2,3-dihydroxybutanedioic acid;ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N068125G View Documents TCI America
N068125G
25g Each for $537.78
 
N06815G View Documents TCI America
N06815G
5g Each for $157.36
 
Specifications

Chemical Identifiers

83602-38-4
307.299
HHPGQKZOPPDLNH-FWQMQHHGSA-N
118855422
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
C12H21NO8
MFCD03093639
(S)-Nipecotic Acid Ethyl Ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, (S)-3-Piperidinecarboxylic Acid Ethyl Ester D-Tartrate
(3S)-2,3-dihydroxybutanedioic acid;ethyl piperidine-3-carboxylate

Specifications

White
25g
C12H21NO8
(S)-Nipecotic Acid Ethyl Ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, (S)-3-Piperidinecarboxylic Acid Ethyl Ester D-Tartrate
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
307.299
307.30
≥98.0% (T)
156°C
83602-38-4
MFCD03093639
HHPGQKZOPPDLNH-FWQMQHHGSA-N
(3S)-2,3-dihydroxybutanedioic acid;ethyl piperidine-3-carboxylate
118855422
Crystal-Powder at 20°C
Ethyl (S)-3-Piperidinecarboxylate D-Tartrate
Safety and Handling

Safety and Handling

TSCA : No