missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (S)-(+)-3-hydroxybutyrate

For synthesis of optically active products

$141.88 - $586.00

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-YFKPBYRVSA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl (3S)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6318403 View Documents Alfa Aesar™
J6318403
1g Each for $141.88
 
AAJ6318406 View Documents Alfa Aesar™
J6318406
5g Each for $586.00
 
Description

Description

Applications
Ethyl (S)-(+)-3-hydroxybutyrate is used for synthesis of optically active products, used as medical and organic intermediates, an important amino protective agents, pharmaceuticals and synthetic organic makes intermediates, organic synthesis used to introduce t-Boc protect gene.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

56816-01-4
132.159
OMSUIQOIVADKIM-YFKPBYRVSA-N
6950306
CCOC(=O)CC(C)O
C6H12O3
MFCD00066206
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
ethyl (3S)-3-hydroxybutanoate

Specifications

Colorless to Yellow
56816-01-4
MFCD00066206
Soluble in chloroform
CCOC(=O)CC(C)O
132.159
132.16
Ethyl (S)-(+)-3-hydroxybutyrate
1g
C6H12O3
s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
OMSUIQOIVADKIM-YFKPBYRVSA-N
ethyl (3S)-3-hydroxybutanoate
6950306
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning