missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (S)-(+)-3-Hydroxybutyrate 96.0+%, TCI America™

$291.31 - $1,010.56

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-YFKPBYRVSA-N
Synonym butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl 3s-3-hydroxybutanoate, ethyl s-+-3-hydroxybutyrate, ethyl s---3-hydroxybutyrate, ethyl s-3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, s-+-3-hydroxybutyric acid ethyl ester, s-ethyl 3-hydroxybutanoate
PubChem CID 6950306
IUPAC Name ethyl (3S)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09755G View Documents Tci America
H09755G
5g Each for $291.31
 
H097525G View Documents Tci America
H097525G
25g Each for $1,010.56
 
Specifications

Chemical Identifiers

56816-01-4
132.159
OMSUIQOIVADKIM-YFKPBYRVSA-N
6950306
CCOC(=O)CC(C)O
C6H12O3
MFCD00066206
butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl 3s-3-hydroxybutanoate, ethyl s-+-3-hydroxybutyrate, ethyl s---3-hydroxybutyrate, ethyl s-3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, s-+-3-hydroxybutyric acid ethyl ester, s-ethyl 3-hydroxybutanoate
ethyl (3S)-3-hydroxybutanoate

Specifications

Yellow
56816-01-4
MFCD00066206
OMSUIQOIVADKIM-YFKPBYRVSA-N
ethyl (3S)-3-hydroxybutanoate
6950306
Clear Liquid at 20°C
Ethyl (S)-(+)-3-Hydroxybutyrate
5g
C6H12O3
butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl 3s-3-hydroxybutanoate, ethyl s-+-3-hydroxybutyrate, ethyl s---3-hydroxybutyrate, ethyl s-3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, s-+-3-hydroxybutyric acid ethyl ester, s-ethyl 3-hydroxybutanoate
CCOC(=O)CC(C)O
132.159
132.16
≥96.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1430

TSCA : No