Please login to your online account to display your discounted pricing

(-)-Ethyl (S)-3-hydroxy-3-phenylpropionate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367910010

 View more versions of this product

Catalog No. AC367910010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 194.23
Quantity 1g
Color Clear colorless
Beilstein 10, III, 540
CAS Max % 100.0
Specific Rotation -49° to -53° (20°C, 589nm) (C=1, CHCl3)
Linear Formula C6H5CH(OH)CH2COOCH2CH3
Molecular Formula C11 H14 O3
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.5090 to 1.5100 (20°C, 589nm)
Chemical Name or Material (-)-Ethyl (S)-3-hydroxy-3-phenylpropionate
Infrared Spectrum Authentic
MDL Number MFCD00216721
CAS 33401-74-0

Specifications

Chemical Name or Material (-)-Ethyl (S)-3-hydroxy-3-phenylpropionate
CAS 33401-74-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11 H14 O3
Linear Formula C6H5CH(OH)CH2COOCH2CH3
Formula Weight 194.23
MDL Number MFCD00216721