missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Ethyl (S)-3-hydroxy-3-phenylpropionate, 98%, ACROS Organics™

Chemical Identifiers

CAS 33401-74-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00216721
InChI Key DVIBDQWVFHDBOP-JTQLQIEISA-N
Synonym --ethyl s-3-hydroxy-3-phenylpropionate
PubChem CID 853697
IUPAC Name ethyl (3S)-3-hydroxy-3-phenylpropanoate
SMILES CCOC(=O)CC(C1=CC=CC=C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367910010 SDS View Documents Thermo Fisher Scientific
AC367910010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

33401-74-0
194.23
DVIBDQWVFHDBOP-JTQLQIEISA-N
853697
CCOC(=O)CC(C1=CC=CC=C1)O
C11H14O3
MFCD00216721
--ethyl s-3-hydroxy-3-phenylpropionate
ethyl (3S)-3-hydroxy-3-phenylpropanoate

Specifications

Authentic
1.5090 to 1.5100 (20°C, 589nm)
Colorless
33401-74-0
100.0
C11H14O3
MFCD00216721
--ethyl s-3-hydroxy-3-phenylpropionate
CCOC(=O)CC(C1=CC=CC=C1)O
194.23
194.23
98%
Glass bottle
-49° to -53° (20°C, 589nm) (C=1, CHCl3)
1g
97.5
97.5% min. (GC)
C6H5CH(OH)CH2COOCH2CH3
10, III, 540
DVIBDQWVFHDBOP-JTQLQIEISA-N
ethyl (3S)-3-hydroxy-3-phenylpropanoate
853697
Liquid
(-)-Ethyl (S)-3-hydroxy-3-phenylpropionate