Please login to your online account to display your discounted pricing

Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate
Name Note 99%
CAS 7149-65-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H11NO3
Formula Weight 157.16
MDL Number MFCD00064497
Synonym 5-Oxo-proline ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC233320100

  SDS
fisher scientific acros
233320100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Low Melting Solid
Formula Weight 157.16
Quantity 10g
Color White to cream
EINECS Number 230-480-9
Specific Rotation Condition - 3.50 (20.00°C c=5,water)
Chemical Name or Material Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate
Specific Rotation -2° to -5° (20°C, 589nm) (c=5, H2O)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H11NO3
MDL Number MFCD00064497
CAS 7149-65-7
Synonym 5-Oxo-proline ethyl ester
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.