Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (S)-2-oxiranylacetate, 97%

$293.82 - $293.82

Chemical Identifiers

Chemical Name or Material Ethyl (S)-2-oxiranylacetate
CAS 112083-63-3
Molecular Formula C6H10O3
Formula Weight 130.14
MDL Number MFCD00672889
Synonym Ethyl (S)-3,4-epoxybutyrate; (S)-2-Oxiraneacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27891ME

  SDS
alfa aesar™
H27891ME
500mg Each for $293.82
Description & Specifications

Specifications

Formula Weight 130.14
Molecular Formula C6H10O3
Quantity 500mg
Synonym Ethyl (S)-3,4-epoxybutyrate; (S)-2-Oxiraneacetic acid ethyl ester
Chemical Name or Material Ethyl (S)-2-oxiranylacetate
Recommended Storage 2° to 8°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
MDL Number MFCD00672889
CAS 112083-63-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%

Applications
Mainly employed inorganic synthesis. They are starting materials for the condensation reactions. Homoallylic alcohol is readily available from the regiospecific ring opening of commercially available ethyl (S)-oxiranyl acetate with the cuprate derived from vinylmagnesium bromide and copper bromide-dimethyl sulfide complex in 88 % yield.

Solubility
Decomposes in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, excess heat, air. Store at 2-8°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C