missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (S)-2-oxiranylacetate, 97%

Manufacturer:  Alfa Aesar™ H27891ME

Catalog No. AAH27891ME



Description

Description

Applications
Mainly employed inorganic synthesis. They are starting materials for the condensation reactions. Homoallylic alcohol is readily available from the regiospecific ring opening of commercially available ethyl (S)-oxiranyl acetate with
Specifications

Specifications

Ethyl (S)-2-oxiranylacetate
500mg
C6H10O3
2-oxiraneacetic acid,ethyl ester, 2s, ethyl 2-2s-oxiran-2-yl acetate, ethyl 2s-oxiran-2-ylacetate, ethyl s-2-oxiranylacetate, ethyl s-3,4-epoxybutanoate, ethyl s-3,4-epoxybutyrate, ethyl s-oxirane-2-acetate, oxiraneacetic acid, ethyl ester, 2s, s-ethyl 2-oxiran-2-yl acetate, s-oxiraneacetic acid ethyl ester
WHUSTVAXKRFVPD-YFKPBYRVSA-N
ethyl 2-[(2S)-oxiran-2-yl]acetate
10920539
97%
Air sensitive
112083-63-3
MFCD00672889
Decomposes in water.
CCOC(=O)CC1CO1
130.143
130.14

Chemical Identifiers

112083-63-3
130.143
WHUSTVAXKRFVPD-YFKPBYRVSA-N
10920539
CCOC(=O)CC1CO1
C6H10O3
MFCD00672889
2-oxiraneacetic acid,ethyl ester, 2s, ethyl 2-2s-oxiran-2-yl acetate, ethyl 2s-oxiran-2-ylacetate, ethyl s-2-oxiranylacetate, ethyl s-3,4-epoxybutanoate, ethyl s-3,4-epoxybutyrate, ethyl s-oxirane-2-acetate, oxiraneacetic acid, ethyl ester, 2s, s-ethyl 2-oxiran-2-yl acetate, s-oxiraneacetic acid ethyl ester
ethyl 2-[(2S)-oxiran-2-yl]acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents