missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (S)-(-)-2-methoxypropionate, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 331730100

 View more versions of this product

Catalog No. AC331730100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to slightly yellow
− 89.00 (20.00°C c=neat)
-86° to -92° (20°C, 589nm) (neat)
Glass bottle
C6H12O3
98%
CH3CH(OCH3)COOCH2CH3
132.16
10mL
03, III, 446
1.3970 to 1.3990 (20°C, 589nm)
Authentic
41918-08-5
Ethyl (S)-(-)-2-methoxypropionate
MFCD01632590

Chemical Identifiers

98%
CH3CH(OCH3)COOCH2CH3
MFCD01632590
≥97.5%
C6H12O3
132.16
Ethyl (S)-(-)-2-methoxypropionate