Please login to your online account to display your discounted pricing

Ethyl (S)-(-)-2-methoxypropionate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl (S)-(-)-2-methoxypropionate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H12O3
Linear Formula CH3CH(OCH3)COOCH2CH3
Formula Weight 132.16
MDL Number MFCD01632590
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC331730010

  SDS
thermo fisher scientific
331730010
Glass Ampule with Screw-cap 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC331730100

  SDS
fisher scientific acros
331730100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 132.16
Quantity 1mL
Color Clear colorless to slightly yellow
Packaging Glass Ampule with Screw-cap
Specific Rotation Condition − 89.00 (20.00°C c=neat)
Beilstein 03, III, 446
Refractive Index 1.3970 to 1.3990 (20°C, 589nm)
Specific Rotation -86° to -92° (20°C, 589nm) (neat)
Infrared Spectrum Authentic
Linear Formula CH3CH(OCH3)COOCH2CH3
MDL Number MFCD01632590
CAS 41918-08-5
Assay Percent Range 98%
Chemical Name or Material Ethyl (S)-(-)-2-methoxypropionate
Molecular Formula C6H12O3