missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (R)-nipecotate L-tartrate, 98%

Manufacturer:  Alfa Aesar™ H5622603

 View more versions of this product

Catalog No. AAH5622603



Specifications

Specifications

Ethyl (R)-nipecotate L-tartrate
157°C to 159°C
167392-57-6
MFCD00799538
HHPGQKZOPPDLNH-RATVZYDNSA-N
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
53249980
98%
+10° (c=5 in Water)
1g
C12H21NO8
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
307.299
307.3

Chemical Identifiers

167392-57-6
307.299
HHPGQKZOPPDLNH-RATVZYDNSA-N
53249980
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
C12H21NO8
MFCD00799538
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Documents