Please login to your online account to display your discounted pricing

Ethyl (R)-nipecotate L-tartarate, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  334780010

 View more versions of this product

Catalog No. AC334780010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 307.3
Quantity 1g
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Molecular Formula C8H15NO2·C4H6O6
MDL Number MFCD00799538
Name Note 97%
Chemical Name or Material Ethyl (R)-nipecotate L-tartarate
CAS 167392-57-6
Assay Percent Range 97%

Specifications

Chemical Name or Material Ethyl (R)-nipecotate L-tartarate
Name Note 97%
CAS 167392-57-6
Assay Percent Range 97%
Molecular Formula C8H15NO2·C4H6O6
Formula Weight 307.3
MDL Number MFCD00799538
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents