Please login to your online account to display your discounted pricing

Ethyl (R)-4-cyano-3-hydroxybutanoate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material ethyle(R)-4-cyano-3-hydroxybutanoate
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7H11NO3
Linear Formula NCCHCH(OH)CHCOOCHCH3
Formula Weight 157.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC348890010

  SDS
thermo fisher scientific
AC348890010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC348890050

  SDS
thermo fisher scientific
AC348890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94%
Formula Weight 157.17
Quantity 1g
Color Colorless to yellow
Density 1.114g/mL
Specific Rotation -4.8° to -5.2° (20°C, 589nm) (neat)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula NCCHCH(OH)CHCOOCHCH3
CAS 141942-85-0
Assay Percent Range 95%
Chemical Name or Material ethyle(R)-4-cyano-3-hydroxybutanoate
Molecular Formula C7H11NO3
Refractive Index 1.4460 to 1.4500 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.