missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 97%, ee 96%

$72.93 - $249.82

Chemical Identifiers

CAS 90866-33-4
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.601
MDL Number MFCD00211242
InChI Key ZAJNMXDBJKCCAT-RXMQYKEDSA-N
Synonym ethyl r-+-4-chloro-3-hydroxybutyrate, r-ethyl 4-chloro-3-hydroxybutanoate, ethyl 3r-4-chloro-3-hydroxybutanoate, ethyl r-4-chloro-3-hydroxybutyrate, ethyl r-+-4-chloro-3-hydroxybutanoate, r-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-butyric acid ethyl ester, r-4-chloro-3-hydroxy-n-butyric acid ethyl ester, ethyl r-4-chloro-3-hydroxybutanoate, butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
PubChem CID 2734445
IUPAC Name ethyl (3R)-4-chloro-3-hydroxybutanoate
SMILES CCOC(=O)CC(CCl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1855303 View Documents Alfa Aesar™
L1855303
1g Each for $72.93
 
AAL1855306 View Documents Alfa Aesar™
L1855306
5g Each for $249.82
 
Specifications

Chemical Identifiers

90866-33-4
166.601
ZAJNMXDBJKCCAT-RXMQYKEDSA-N
2734445
CCOC(=O)CC(CCl)O
C6H11ClO3
MFCD00211242
ethyl r-+-4-chloro-3-hydroxybutyrate, r-ethyl 4-chloro-3-hydroxybutanoate, ethyl 3r-4-chloro-3-hydroxybutanoate, ethyl r-4-chloro-3-hydroxybutyrate, ethyl r-+-4-chloro-3-hydroxybutanoate, r-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-butyric acid ethyl ester, r-4-chloro-3-hydroxy-n-butyric acid ethyl ester, ethyl r-4-chloro-3-hydroxybutanoate, butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
ethyl (3R)-4-chloro-3-hydroxybutanoate

Specifications

>110°C (230°F)
93°C to 95°C (5mmHg)
90866-33-4
MFCD00211242
ZAJNMXDBJKCCAT-RXMQYKEDSA-N
ethyl (3R)-4-chloro-3-hydroxybutanoate
2734445
97%
ee 96%
+14° (Neat)
1g
C6H11ClO3
ethyl r-+-4-chloro-3-hydroxybutyrate, r-ethyl 4-chloro-3-hydroxybutanoate, ethyl 3r-4-chloro-3-hydroxybutanoate, ethyl r-4-chloro-3-hydroxybutyrate, ethyl r-+-4-chloro-3-hydroxybutanoate, r-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-butyric acid ethyl ester, r-4-chloro-3-hydroxy-n-butyric acid ethyl ester, ethyl r-4-chloro-3-hydroxybutanoate, butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
CCOC(=O)CC(CCl)O
166.601
166.61
Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H312-H332-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

Danger

Recommended Storage : 2° to 8°C