missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (R)-(-)-3-Piperidinecarboxylate, TCI America™

$175.70 - $524.03

Chemical Identifiers

CAS 25137-01-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD03093637
InChI Key XIWBSOUNZWSFKU-SSDOTTSWSA-N
Synonym r-ethyl piperidine-3-carboxylate, ethyl 3r-piperidine-3-carboxylate, ethyl r-nipecotate, r-ethyl nipecotate, r-nipecotic acid ethyl ester, r---nipecotic acid ethyl ester, ethyl 3r-piperidine-3-carboxylate, r---ethyl nipecotate, r-piperidine-3-carboxylic acid ethyl ester, ethyl r---nipecotate
PubChem CID 185582
IUPAC Name ethyl (3R)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06555G View Documents TCI America
N06555G
5g Each for $175.70
 
N065525G View Documents TCI America
N065525G
25g Each for $524.03
 
Specifications

Chemical Identifiers

25137-01-3
157.213
XIWBSOUNZWSFKU-SSDOTTSWSA-N
185582
CCOC(=O)C1CCCNC1
C8H15NO2
MFCD03093637
r-ethyl piperidine-3-carboxylate, ethyl 3r-piperidine-3-carboxylate, ethyl r-nipecotate, r-ethyl nipecotate, r-nipecotic acid ethyl ester, r---nipecotic acid ethyl ester, ethyl 3r-piperidine-3-carboxylate, r---ethyl nipecotate, r-piperidine-3-carboxylic acid ethyl ester, ethyl r---nipecotate
ethyl (3R)-piperidine-3-carboxylate

Specifications

110°C
5g
C8H15NO2
r-ethyl piperidine-3-carboxylate, ethyl 3r-piperidine-3-carboxylate, ethyl r-nipecotate, r-ethyl nipecotate, r-nipecotic acid ethyl ester, r---nipecotic acid ethyl ester, ethyl 3r-piperidine-3-carboxylate, r---ethyl nipecotate, r-piperidine-3-carboxylic acid ethyl ester, ethyl r---nipecotate
CCOC(=O)C1CCCNC1
157.213
157.21
≥98.0% (GC)
Yellow
25137-01-3
MFCD03093637
XIWBSOUNZWSFKU-SSDOTTSWSA-N
ethyl (3R)-piperidine-3-carboxylate
185582
Clear Liquid at 20°C
Ethyl (R)-(-)-3-Piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator