missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (R)-3-Piperidinecarboxylate L-Tartrate 98.0+%, TCI America™

Manufacturer:  TCI America N068025G

 View more versions of this product

Catalog No. N068025G



Specifications

Specifications

Ethyl (R)-3-Piperidinecarboxylate L-Tartrate
157°C
167392-57-6
MFCD00799538
HHPGQKZOPPDLNH-RATVZYDNSA-N
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
53249980
Crystal-Powder at 20°C
White
25g
C12H21NO8
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
307.299
307.30
≥98.0% (T)

Chemical Identifiers

167392-57-6
307.299
HHPGQKZOPPDLNH-RATVZYDNSA-N
53249980
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
C12H21NO8
MFCD00799538
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents