missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl (R)-3-hydroxybutyrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 345760010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl (R)-3-hydroxybutyrate | |
24915-95-5 | |
75°C to 76°C (12.0mmHg) | |
97.5% min. (GC) | |
C6H12O3 | |
CH3CH(OH)CH2COOCH2CH3 | |
1g | |
−45.5° (20°C 589nm,c=1,CHCl3) | |
ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester | |
OMSUIQOIVADKIM-RXMQYKEDSA-N | |
ethyl (3R)-3-hydroxybutanoate | |
440030 | |
132.16 |
98% | |
1.0170g/mL | |
Authentic | |
Glass bottle | |
1.4195 to 1.4215 | |
MFCD00075386 | |
1.017 | |
-45.5 | |
Solubility in water: soluble in water | |
CCOC(=O)CC(C)O | |
132.16 | |
CHEBI:28707 | |
98% |
Chemical Identifiers
24915-95-5 | |
132.16 | |
OMSUIQOIVADKIM-RXMQYKEDSA-N | |
440030 | |
ethyl (3R)-3-hydroxybutanoate |
C6H12O3 | |
MFCD00075386 | |
ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester | |
CHEBI:28707 | |
CCOC(=O)CC(C)O |
RUO â Research Use Only